Anti-Digoxin Mouse monoclonal antibody unconjugated
Lieferant: QED Bioscience
19102
QEDB19102EA 405 EUR
QEDB19102
Anti-Digoxin Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Digoxin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen