Anti-Endothelin Mouse monoclonal antibody unconjugated
Lieferant: QED Bioscience
19404
QEDB19404EA 499 EUR
QEDB19404
Anti-Endothelin Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Endothelin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human