Anti-Influenza B Neuraminidase Mouse Monoclonal Antibody
Lieferant: QED Bioscience
19602
QEDB19602EA 587 EUR
QEDB19602
Anti-Influenza B Neuraminidase Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Influenza B Neuraminidase
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestellen