Anti-SAP30L Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
28113
QEDB28113EA 391 EUR
QEDB28113
Anti-SAP30L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SAP30L (SAP30-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR