Anti-UBIAD1 Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
34031
QEDB34031EA 283 EUR
QEDB34031
Anti-UBIAD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UBIAD1 (UbiA prenyltransferase domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR