Anti-Interleukin-1 Receptor (IL-1R) Mouse Monoclonal Antibody

Lieferant: QED Bioscience
40101
QEDB40101EA 385 EUR
QEDB40101
Anti-Interleukin-1 Receptor (IL-1R) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Interleukin-1 Receptor (IL-1R)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR