Anti-CALR Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
56259
QEDB56259EA 319 EUR
QEDB56259
Anti-CALR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CALR (calreticulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR