Anti-Piccolo Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
56496
QEDB56496EA 482 EUR
QEDB56496
Anti-Piccolo Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Piccolo
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR