Anti-TATA Binding Protein Mouse Monoclonal Antibody
Lieferant: QED Bioscience
70102
QEDB70102EA 424 EUR
QEDB70102
Anti-TATA Binding Protein Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TATA Binding Protein
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat