Bulk adsorbents, Ottawa sand
Lieferant: Restek
26137
REST26137EA 412 EUR
REST26137
Bulk adsorbents, Ottawa sand
Chromatographie-Sorbentien SPE-Sorbentien
Ottawa sand is listed in several U.S. EPA methods as the specified medium for matrix spike and laboratory control blanks.
Verpackung: Packaged in convenient 5 kg buckets.