IgG2a kappa Isotype Control (Unconjugated)

Lieferant: Rockland Immunochemicals
012-001-332
ROCK012-001-332EA 400 EUR
ROCK012-001-332
IgG2a kappa Isotype Control (Unconjugated)
Antikörper
Rat IgG2a Kappa isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.

Recommended Dilutions: Flow Cytometry: 1:1000-1:5000;

Type: Control
Antigen: IgG2a κ
Clonality: Monoclonal
Clone: RG2aK
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR