IgG2b kappa Isotype Control (Unconjugated)
Lieferant: Rockland Immunochemicals
012-001-334
ROCK012-001-334EA 400 EUR
ROCK012-001-334
IgG2b kappa Isotype Control (Unconjugated)
Antikörper
Rat IgG2b Kappa isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.
Recommended Dilutions: Flow Cytometry: 1:1000-1:5000;
Type: Control
Antigen: IgG2b κ
Clonality: Monoclonal
Clone: RG2bK
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity:
Bestellen