Anti-Bilirubin Oxidase Goat Polyclonal Antibody
Lieferant: Rockland Immunochemicals
100-101-212
ROCK100-101-212EA 2060 EUR
ROCK100-101-212
Anti-Bilirubin Oxidase Goat Polyclonal Antibody
Antikörper
Primary Goat Anti-BILIRUBIN OXIDASE (Myrothecium verrucaria) Reacts with Pan
Recommended Dilutions: ELISA: 1:25000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: Bilirubin Oxidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Myrothecium Verrucaria