Anti-ALDOA Goat Polyclonal Antibody
Lieferant: Rockland Immunochemicals
100-1141
ROCK100-1141EA 2060 EUR
ROCK100-1141
Anti-ALDOA Goat Polyclonal Antibody
Antikörper
This product has been assayed against 1.0 µg of Aldolase (Rabbit Muscle) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG (H&L) (Rabbit) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: ALDOA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit