Anti-NOTCH2 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
100-401-408
ROCK100-401-408EA 821 EUR
ROCK100-401-408
Anti-NOTCH2 Rabbit Polyclonal Antibody
Antikörper
Anti-Notch2 has a strong response was detected by ELISA against the immunizing peptide.This product was assayed against the peptide immunogen in a standard capture ELISA using Peroxidase conjugated anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:400 - 1:2000; Immunochemistry: 1:500;

Type: Primary
Antigen: NOTCH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR