Anti-PIK3CD Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
100-401-862
ROCK100-401-862EA 821 EUR
ROCK100-401-862
Anti-PIK3CD Rabbit Polyclonal Antibody
Antikörper
This antibody has been tested for use in ELISA, western blotting and immunoprecipitation.  Reactivity in other immunoassays is unknown.  A mouse whole cell splenic lysate is suitable for use as a positive control.
Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:5000;
Type: Primary
Antigen: PIK3CD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Human