Anti-GADD45G Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
100-401-863
ROCK100-401-863EA 821 EUR
ROCK100-401-863
Anti-GADD45G Rabbit Polyclonal Antibody
Antikörper
This antibody has been tested for use in ELISA, western blotting and immunoprecipitation.  Reactivity in other immunoassays is unknown. This product was assayed by western blotting after immunoprecipitation of mouse GADD45 gamma from a lysate.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:100 - 1:500; Immunoprecipitation: Use 10 µl;

Type: Primary
Antigen: GADD45G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR