Anti-CUL7 Rabbit Monoclonal Antibody (Unconjugated) [clone: 18E1]

Lieferant: Rockland Immunochemicals
100-401-A06
ROCK100-401-A06EA 821 EUR
ROCK100-401-A06
Anti-CUL7 Rabbit Monoclonal Antibody (Unconjugated) [clone: 18E1]
Antikörper
This antibody reacts with human and mouse Cul7 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated Protein and also immunoprecipitates Protein from cell lysates (using HeLa, NIH-3T3, and others).

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: CUL7
Clonality: Monoclonal
Clone: 18E1
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR