Anti-SKP1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
100-401-A08
ROCK100-401-A08EA 821 EUR
ROCK100-401-A08
Anti-SKP1 Rabbit Polyclonal Antibody
Antikörper
This antibody reacts with human SKP1 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated Protein and Protein from cell lysates (using HeLa, NIH-3T3, and others).

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: SKP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR