Anti-TICAM1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
100-401-I35
ROCK100-401-I35EA 939 EUR
ROCK100-401-I35
Anti-TICAM1 Rabbit Polyclonal Antibody
Antikörper
Anti-TRIF antibody is sutibale for use in WB. Expect a band approximately 76kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: TICAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR