Anti-MYO6 Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
100-401-J03S
ROCK100-401-J03SEA 226 EUR
ROCK100-401-J03S
Anti-MYO6 Rabbit Polyclonal Antibody
Antikörper
Myosin-6 antibody has been tested for use in ELISA and western blot. For western blots expect a band of approximately 150 kDa in size corresponding to truncated myosin-6 protein. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000
Type: Primary
Antigen: MYO6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human