Anti-ALDH Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
100-4144
ROCK100-4144EA 2060 EUR
ROCK100-4144
Anti-ALDH Rabbit Polyclonal Antibody
Antikörper
This product has been assayed against 1.0 µg of Aldehyde Dehydrogenase (Yeast) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: ALDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast