Anti-Penicillinase Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
100-4175
ROCK100-4175EA 2060 EUR
ROCK100-4175
Anti-Penicillinase Rabbit Polyclonal Antibody
Antikörper
Anti-PENICILLINASE Antibody has been assayed against 1.0 µg of Penicillinase (Enterobacter cloacae) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Penicillinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Enterobacter cloacae
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR