Anti-KTI3 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
100-4179
ROCK100-4179EA 2060 EUR
ROCK100-4179
Anti-KTI3 Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-TRYPSIN INHIBITOR (Soy Bean) Reacts with Pan

Recommended Dilutions: ELISA: 1:3000 - 1:15000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: KTI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Soy Bean
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR