Anti-MDH1 Sheep Polyclonal Antibody
Lieferant: Rockland Immunochemicals
100-601-145
ROCK100-601-145EA 2060 EUR
ROCK100-601-145
Anti-MDH1 Sheep Polyclonal Antibody
Antikörper
This product has been assayed against 1.0 µg of Malate Dehydrogenase (Pig Heart) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Sheep IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: MDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Pig