Anti-Mouse Serum Rabbit Monoclonal Antibody (Unconjugated) [clone: 2D11.H5]

Lieferant: Rockland Immunochemicals
110-4101
ROCK110-4101EA 504 EUR
ROCK110-4101
Anti-Mouse Serum Rabbit Monoclonal Antibody (Unconjugated) [clone: 2D11.H5]
Antikörper
Anti-MOUSE SERUM (RABBIT) Antibody is suitable for immunoprecipitation, immunodiffusion, conjugation and most immunological methods requiring high titer and specificity.

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Mouse Serum
Clonality: Monoclonal
Clone: 2D11.H5
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR