Anti-IgG Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Rockland Immunochemicals
18-0216-32
ROCK18-0216-32EA 457 EUR
ROCK18-0216-32
Anti-IgG Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Fluorescent Rabbit TrueBlot® Antibody Fluorescein is provided as a lyophilized powder. In order to conserve reagent, we recommend incubating the blots from minigels in sealed bags (removing as much air as possible) with minimal volume (2-3 mls).

Recommended Dilutions: Restore with deionised water (or equivalent); 100 µl.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: eB182
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR