Anti-CHOX_ALCSP Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-101-094S
ROCK200-101-094SEA 226 EUR
ROCK200-101-094S
Anti-CHOX_ALCSP Goat Polyclonal Antibody
Antikörper
CHOLINE OXIDASE Antibody has been assayed against 1.0 ug of Choline Oxidase [Alcaligenes species] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG [H&L] (Rabbit) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100; Other Dilution: User Optimized

Type: Primary
Antigen: CHOX_ALCSP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Alcaligenes
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR