Anti-RFP Polyclonal Antibody
Lieferant: Rockland Immunochemicals
200-101-379S
ROCK200-101-379SEA 226 EUR
ROCK200-101-379S
Anti-RFP Polyclonal Antibody
Antikörper
Goat anti-red fluorecent protein antibody
  • Purified IgG Fraction
  • No reaction was observed against human, mouse or rat serum proteins
  • Western blot
Antibodies to RFP (Discosoma spp.) are intended for use in immunological assays including ELISA, western blotting, immunofluorescence, and fluorescence activated cell sorting (FACS).
Polyclonal anti-RFP is designed to detect RFP and its variants. Optimal titers for applications should be determined by the researcher. Expect a band ~27kDa in Western Blot in the appropriate cell lysate or extract.