Anti-GST Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-101-C45S
ROCK200-101-C45SEA 226 EUR
ROCK200-101-C45S
Anti-GST Goat Polyclonal Antibody
Antikörper
Suitable for immunoblotting (western or dot blot), ELISA, immunoprecipitation and most immunological methods requiring high titer and specificity.

Recommended Dilutions: ELISA: 1:30,000 - 1:100000; Western Blot: 1:3000 - 1:20000;

Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR