Anti-CDK1 Mouse Polyclonal Antibody (Unconjugated) [clone: POH-1]

Lieferant: Rockland Immunochemicals
200-301-160
ROCK200-301-160EA 841 EUR
ROCK200-301-160
Anti-CDK1 Mouse Polyclonal Antibody (Unconjugated) [clone: POH-1]
Antikörper
Primary Mouse Anti-cdc2 (p34) Reacts with Human, Mouse, Rat

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDK1
Clonality: Polyclonal
Clone: POH-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR