Anti-CDKN1A Mouse Polyclonal Antibody (Unconjugated) [clone: WA-1]

Lieferant: Rockland Immunochemicals
200-301-171
ROCK200-301-171EA 841 EUR
ROCK200-301-171
Anti-CDKN1A Mouse Polyclonal Antibody (Unconjugated) [clone: WA-1]
Antikörper
Primary Mouse Anti-p21/WAF1 Reacts with Human

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:2000 - 1:10000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDKN1A
Clonality: Polyclonal
Clone: WA-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR