Anti-AKT1 Mouse Polyclonal Antibody (Unconjugated) [clone: 18F3.H11]

Lieferant: Rockland Immunochemicals
200-301-269S
ROCK200-301-269SEA 226 EUR
ROCK200-301-269S
Anti-AKT1 Mouse Polyclonal Antibody (Unconjugated) [clone: 18F3.H11]
Antikörper
Anti-AKT pT308 antibody is suitable for ELISA, immunochemistry, immunoprecipitation and western blotting.  Expect a band approximately 56 kDa in size corresponding to phosphorylated AKT Protein by western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:500 - 1:3000; Immunochemistry: 20 µg/ml;

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone: 18F3.H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human. Mouse, Rat, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR