Anti-IDO1 Mouse Monoclonal Antibody (Unconjugated) [clone: 10.1]

Lieferant: Rockland Immunochemicals
200-301-E57S
ROCK200-301-E57SEA 226 EUR
ROCK200-301-E57S
Anti-IDO1 Mouse Monoclonal Antibody (Unconjugated) [clone: 10.1]
Antikörper
Type: Primary
Antigen: IDO1
Clonality: Monoclonal
Clone: 10.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR