Anti-CRYAA Mouse Monoclonal Antibody (Unconjugated) [clone: 1H3.B8]

Lieferant: Rockland Immunochemicals
200-301-E84
ROCK200-301-E84EA 841 EUR
ROCK200-301-E84
Anti-CRYAA Mouse Monoclonal Antibody (Unconjugated) [clone: 1H3.B8]
Antikörper
Anti-Alpha A Crystallin Antibody is recommended for use in WB and ELISA. Expect a band approximately ~20kDa Protein corresponding to the molecular mass of a-crystallin on SDS PAGE immunoblots.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:2000

Type: Primary
Antigen: CRYAA
Clonality: Monoclonal
Clone: 1H3.B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Bovine, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR