Anti-ERD2 Mouse Monoclonal Antibody (Unconjugated) [clone: 2E7]
Lieferant: Rockland Immunochemicals
200-301-F44
ROCK200-301-F44EA 841 EUR
ROCK200-301-F44
Anti-ERD2 Mouse Monoclonal Antibody (Unconjugated) [clone: 2E7]
Antikörper
Anti-HDEL Antibody is recommended for use in WB and IF microscopy. Expect a band approximately ~78kDa corresponding to specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: Immunofluorescence: 1:50-1:500; Western Blot: 1:1000-1:2000
Type: Primary
Antigen: ERD2
Clonality: Monoclonal
Clone: 2E7
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG2b
Reactivity: Drosophilia, Yeast, Saccharomyce, Barnyard Grass, Beet, Cotton, Mung Bean, Sorghum, Wheat, Yeast