Anti-BAZ1B Mouse Monoclonal Antibody (Unconjugated) [clone: H9010]
Lieferant: Rockland Immunochemicals
200-301-F66
ROCK200-301-F66EA 841 EUR
ROCK200-301-F66
Anti-BAZ1B Mouse Monoclonal Antibody (Unconjugated) [clone: H9010]
Antikörper
Anti-Hsp90 Antibody is suitable for WB, IP, ELISA, and IHC. Expect a band approximately 90kD Proteins corresponding to the molecular mass of Hsp90 on SDS Page immunoblots. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:200; Western Blot: 1:500; Immunoprecipitation: 5µg/20µl
Type: Primary
Antigen: BAZ1B
Clonality: Monoclonal
Clone: H9010
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rabbit, Chicken