Anti-HSP90AB1 Mouse Monoclonal Antibody (Unconjugated) [clone: D7A]
Lieferant: Rockland Immunochemicals
200-301-F68
ROCK200-301-F68EA 841 EUR
ROCK200-301-F68
Anti-HSP90AB1 Mouse Monoclonal Antibody (Unconjugated) [clone: D7A]
Antikörper
Anti-Hsp90 Antibody is suitable for WB, IP, ELISA, and IHC. Expect a band approximately 90kD Proteins corresponding to the molecular mass of Hsp90 on SDS Page immunoblots. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:200; Western Blot: 1:500; Immunoprecipitation: 5µg/20µl
Type: Primary
Antigen: HSP90AB1
Clonality: Monoclonal
Clone: D7A
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG
Reactivity: Bovine, Chicken, Human, Mouse, Pig, Rat, Rabbit