Anti-SCN2A Mouse Monoclonal Antibody (Unconjugated) [clone: S69-3]

Lieferant: Rockland Immunochemicals
200-301-G19
ROCK200-301-G19EA 841 EUR
ROCK200-301-G19
Anti-SCN2A Mouse Monoclonal Antibody (Unconjugated) [clone: S69-3]
Antikörper
Anti-Nav1.2 Antibody is suitable for use in WB, IP, and IHC. Expect a band approximately ~250kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: SCN2A
Clonality: Monoclonal
Clone: S69-3
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR