Anti-PCLO Mouse Monoclonal Antibody (Unconjugated) [clone: 6H9-B6]

Lieferant: Rockland Immunochemicals
200-301-G30
ROCK200-301-G30EA 841 EUR
ROCK200-301-G30
Anti-PCLO Mouse Monoclonal Antibody (Unconjugated) [clone: 6H9-B6]
Antikörper
Anti-PIF microscopyolo Antibody is suitable for use in WB and ELISA. Expect a band approximately ~550kDa corresponding to specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200

Type: Primary
Antigen: PCLO
Clonality: Monoclonal
Clone: 6H9-B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR