Anti-TMBIM6 Mouse Monoclonal Antibody (Unconjugated) [clone: 20F430]

Lieferant: Rockland Immunochemicals
200-301-H60
ROCK200-301-H60EA 913 EUR
ROCK200-301-H60
Anti-TMBIM6 Mouse Monoclonal Antibody (Unconjugated) [clone: 20F430]
Antikörper
Anti-Bi-1 antibody is suitable for use in IHC (p). Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 5-10 µg/ml

Type: Primary
Antigen: TMBIM6
Clonality: Monoclonal
Clone: 20F430
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR