Anti-CASP1 Mouse Monoclonal Antibody (Unconjugated) [clone: 14F468]

Lieferant: Rockland Immunochemicals
200-301-H62
ROCK200-301-H62EA 1030 EUR
ROCK200-301-H62
Anti-CASP1 Mouse Monoclonal Antibody (Unconjugated) [clone: 14F468]
Antikörper
Anti-Caspase-1 antibody is suitable for use in IHC (p) and WB. Expect a band approximately 46kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml; Immunochemistry: titrate to determine concentration

Type: Primary
Antigen: CASP1
Clonality: Monoclonal
Clone: 14F468
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR