Anti-DNMT3A Mouse Monoclonal Antibody (Unconjugated) [clone: 64B1446]

Lieferant: Rockland Immunochemicals
200-301-H70
ROCK200-301-H70EA 1050 EUR
ROCK200-301-H70
Anti-DNMT3A Mouse Monoclonal Antibody (Unconjugated) [clone: 64B1446]
Antikörper
Anti-Dnmt3a Antibody is suitable for use in ChIP, IF/ICC, IHC (p) and WB. Expect a band approximately 120 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Immunofluorescence: 5 µg/ml; Western Blot: 2 µg/ml

Type: Primary
Antigen: DNMT3A
Clonality: Monoclonal
Clone: 64B1446
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR