Anti-IKBKG Mouse Monoclonal Antibody (Unconjugated) [clone: 72C627]
Lieferant: Rockland Immunochemicals
200-301-H87
ROCK200-301-H87EA 913 EUR
ROCK200-301-H87
Anti-IKBKG Mouse Monoclonal Antibody (Unconjugated) [clone: 72C627]
Antikörper
Anti-IKKgamma antibody is suitbale for use in Flow (I) and WB. Expect a band approximately 48kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: Flow Cytometry: 0.1-0.5/10^6 cells; Western Blot: 2 µg/ml
Type: Primary
Antigen: IKBKG
Clonality: Monoclonal
Clone: 72C627
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse