Anti-BIRC7 Mouse Monoclonal Antibody (Unconjugated) [clone: 88C570]

Lieferant: Rockland Immunochemicals
200-301-H90
ROCK200-301-H90EA 913 EUR
ROCK200-301-H90
Anti-BIRC7 Mouse Monoclonal Antibody (Unconjugated) [clone: 88C570]
Antikörper
Anti-Livin antibody is suitbale for use in IF/ICC and WB. Expect a band approximately 30kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Immunofluorescence: 2-5 µg/ml; Western Blot: 1-2 µg/ml

Type: Primary
Antigen: BIRC7
Clonality: Monoclonal
Clone: 88C570
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR