Anti-TNFSF11 Mouse Monoclonal Antibody (Unconjugated) [clone: 12A668]

Lieferant: Rockland Immunochemicals
200-301-I07
ROCK200-301-I07EA 957 EUR
ROCK200-301-I07
Anti-TNFSF11 Mouse Monoclonal Antibody (Unconjugated) [clone: 12A668]
Antikörper
Anti-RANKL Antibody is suitable for use in IHC (paraffin) and WB. Expect a band approximately 35.5 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunochemistry: 1:10-1:2000 Paraffin: 5 ug/ml

Type: Primary
Antigen: TNFSF11
Clonality: Monoclonal
Clone: 12A668
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR