Anti-TLR8 Mouse Monoclonal Antibody (Unconjugated) [clone: 44C143]
Lieferant: Rockland Immunochemicals
200-301-I30
ROCK200-301-I30EA 957 EUR
ROCK200-301-I30
Anti-TLR8 Mouse Monoclonal Antibody (Unconjugated) [clone: 44C143]
Antikörper
Anti-TLR8 Antibody is suitable for use in Flow (CS), Flow (I), IHC (p) and WB.Expect a band approximately 121kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: Western Blot: 1-3 µg/ml; Immunochemistry: 5 µg/ml
Type: Primary
Antigen: TLR8
Clonality: Monoclonal
Clone: 44C143
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human