Anti-TERF2 Mouse Monoclonal Antibody (Unconjugated) [clone: 4A794.15]
Lieferant: Rockland Immunochemicals
200-301-I34
ROCK200-301-I34EA 1050 EUR
ROCK200-301-I34
Anti-TERF2 Mouse Monoclonal Antibody (Unconjugated) [clone: 4A794.15]
Antikörper
Anti-TRF2 Antibody is suitable for use in ChIP, ELISA, Flow (Intracellular) Cytometry, IF/ICC, IHC (frozen), IP, and WB. Expect a band approximately 55.6 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 2 µg/ml; ChIP: User Optimized; Immunofluorescence: User Optimized; Western Blot: 2-4 µg/ml; Immunochemistry: 5 µg/ml; Immunoprecipitation: 2 µg/10^6 cells
Type: Primary
Antigen: TERF2
Clonality: Monoclonal
Clone: 4A794.15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human