Anti-CALCA Mouse Monoclonal Antibody (Unconjugated) [clone: 6C12.A2.H4.A2.F9]

Lieferant: Rockland Immunochemicals
200-301-I43
ROCK200-301-I43EA 841 EUR
ROCK200-301-I43
Anti-CALCA Mouse Monoclonal Antibody (Unconjugated) [clone: 6C12.A2.H4.A2.F9]
Antikörper
Anti-Procalcitonin antibody is suitbale for use in ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:1000-1:20000; Western Blot: 1µg/ml;

Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: 6C12.A2.H4.A2.F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR