Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 549) [clone: 17F6.B11]

Lieferant: Rockland Immunochemicals
200-342-268
ROCK200-342-268EA 808 EUR
ROCK200-342-268
Anti-AKT1 Mouse Monoclonal Antibody (DyLight® 549) [clone: 17F6.B11]
Antikörper
This monoclonal antibody is suitable for immunofluorescence microscopy, FLISA and western blotting. Expect a band approximately 56 kDa in size corresponding to phosphorylated AKT Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:4000 - 1:20000; FLISA: >1:20000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 17F6.B11
Conjugation: DyLight® 549
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR